siRNA Details
Gene Symbol: CCDC144A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025429.1 | XP_016880918.1 | 16 | 4754 | ||
GenBank mRNA | AB011137.2 | - | 6 | 2999 | ||
AK125971.1 | - | 12 | 3110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
