siRNA Details
Gene Symbol: NUDCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145266.4 | NP_660309.1 | 3 | 345 | ||
GenBank mRNA | AK311957.1 | - | 3 | 308 | ||
BC017934.1 | - | 3 | 345 | 4698187 | ||
BX538290.1 | - | 3 | 346 | |||
FJ215684.1 | - | 3 | 285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
