siRNA Details
Gene Symbol: DPY19L2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003561.2 | 5 | 503 | |||
NR_027768.1 | 3 | 798 | ||||
GenBank mRNA | AK097711.1 | - | 3 | 726 | ||
AK131353.1 | - | 5 | 496 | |||
AK302046.1 | - | 4 | 711 | |||
AL834175.1 | - | 3 | 551 | |||
BC023575.2 | - | 4 | 900 | |||
BC068601.1 | - | 3 | 609 | |||
BC150309.1 | - | 3 | 259 | |||
BX648212.1 | - | 4 | 780 | |||
DB094748.1 | - | 4 | 402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
