siRNA Details
Gene Symbol: DPY19L2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003561.2 | 5, 6 | 1924 | |||
NR_027768.1 | 3, 4 | 2219 | ||||
GenBank mRNA | AK131353.1 | - | 5, 6 | 1917 | ||
AL834175.1 | - | 3, 4 | 1972 | |||
BC150309.1 | - | 3, 4 | 1680 | |||
BX648212.1 | - | 4, 5 | 2201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
