siRNA Details
Gene Symbol: DPY19L2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003561.2 | 21 | 3504 | |||
NR_027768.1 | 23 | 4165 | ||||
GenBank mRNA | AK093485.1 | - | 17 | 1577 | ||
AL834175.1 | - | 23 | 3918 | |||
BC150309.1 | - | 22 | 3557 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
