siRNA Details
Gene Symbol: SYT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175733.3 | NP_783860.1 | 2 | 424 | ||
XM_011519900.2 | XP_011518202.1 | 2 | 402 | |||
XM_011519901.2 | XP_011518203.1 | 2 | 402 | |||
XM_011519902.2 | XP_011518204.1 | 2 | 175 | |||
XM_011519904.2 | XP_011518206.1 | 2 | 402 | |||
XM_011519906.2 | XP_011518208.1 | 2 | 402 | |||
XM_011519907.2 | XP_011518209.1 | 2 | 402 | |||
GenBank mRNA | AK055003.1 | - | 2 | 204 | ||
AK315013.1 | - | 2 | 367 | |||
BC029605.1 | - | 2 | 181 | 5296925 | ||
BC046367.1 | - | 2 | 424 | 5770087 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
