siRNA Details
Gene Symbol: PSG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031850.3 | NP_001027020.1 | 4 | 926 | ||
NM_002782.4 | NP_002773.1 | 4 | 926 | |||
GenBank mRNA | BC020652.1 | - | 4 | 893 | 4770562 | |
M31125.1 | - | 4 | 839 | |||
M33666.1 | - | 4 | 846 | |||
X17098.1 | - | 4 | 847 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
