siRNA Details
Gene Symbol: BAHCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291324.1 | NP_001278253.1 | 11 | 3334 | ||
XM_011525063.2 | XP_011523365.1 | 12 | 3726 | |||
XM_017024898.1 | XP_016880387.1 | 11 | 3385 | |||
XM_017024899.1 | XP_016880388.1 | 10 | 3291 | |||
GenBank mRNA | AB040880.1 | - | 7 | 1085 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
