siRNA Details
Gene Symbol: JMJD1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282948.1 | NP_001269877.1 | 14 | 6025 | ||
NM_001318153.1 | NP_001305082.1 | 14 | 6029 | |||
NM_001318154.1 | NP_001305083.1 | 15 | 5967 | |||
NM_001322252.1 | NP_001309181.1 | 14 | 6022 | |||
NM_001322254.1 | NP_001309183.1 | 13 | 5919 | |||
NM_001322258.1 | NP_001309187.1 | 14 | 5861 | |||
NM_032776.2 | NP_116165.1 | 15 | 6136 | |||
NR_134512.1 | 11 | 5603 | ||||
XM_011539503.1 | XP_011537805.1 | 14 | 5629 | |||
XM_017015897.1 | XP_016871386.1 | 14 | 5624 | |||
XM_017015898.1 | XP_016871387.1 | 15 | 5817 | |||
XM_017015899.1 | XP_016871388.1 | 15 | 5717 | |||
XM_017015900.1 | XP_016871389.1 | 15 | 6121 | |||
XM_017015901.1 | XP_016871390.1 | 14 | 5664 | |||
XM_017015902.1 | XP_016871391.1 | 14 | 5925 | |||
XM_017015903.1 | XP_016871392.1 | 13 | 5473 | |||
GenBank mRNA | AB037801.1 | - | 6 | 1976 | ||
AK024991.1 | - | 4 | 1437 | |||
AK056660.1 | - | 6 | 2512 | |||
AK095584.1 | - | 8 | 3132 | |||
AL831917.1 | - | 8 | 4664 | |||
BC143722.1 | - | 14 | 5868 | 9052236 | ||
BX537954.1 | - | 11 | 5566 | |||
EF068222.1 | - | 17 | 6093 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
