siRNA Details
Gene Symbol: JMJD1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282948.1 | NP_001269877.1 | 17 | 6752 | ||
NM_001318153.1 | NP_001305082.1 | 17 | 6756 | |||
NM_001318154.1 | NP_001305083.1 | 18 | 6694 | |||
NM_001322252.1 | NP_001309181.1 | 17 | 6749 | |||
NM_001322254.1 | NP_001309183.1 | 16 | 6646 | |||
NM_001322258.1 | NP_001309187.1 | 17 | 6588 | |||
NM_032776.2 | NP_116165.1 | 18 | 6863 | |||
NR_134512.1 | 14 | 6276 | ||||
XM_011539503.1 | XP_011537805.1 | 17 | 6356 | |||
XM_017015897.1 | XP_016871386.1 | 17 | 6351 | |||
XM_017015898.1 | XP_016871387.1 | 18 | 6544 | |||
XM_017015899.1 | XP_016871388.1 | 18 | 6444 | |||
XM_017015900.1 | XP_016871389.1 | 18 | 6848 | |||
XM_017015901.1 | XP_016871390.1 | 17 | 6391 | |||
XM_017015902.1 | XP_016871391.1 | 17 | 6652 | |||
XM_017015903.1 | XP_016871392.1 | 16 | 6200 | |||
GenBank mRNA | AB037801.1 | - | 9 | 2700 | ||
AK024991.1 | - | 7 | 2163 | |||
AL831917.1 | - | 11 | 5391 | |||
BC143722.1 | - | 17 | 6595 | 9052236 | ||
BX537954.1 | - | 14 | 6239 | |||
DR762886.1 | - | 1 | 150 | |||
EF068222.1 | - | 20 | 6820 | |||
L40411.1 | - | 2 | 436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
