siRNA Details
Gene Symbol: ANKRD20A8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003366.2 | 14 | 2282 | |||
GenBank mRNA | CR936649.1 | - | 2 | 3138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
