siRNA Details
Gene Symbol: SCRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033129.3 | NP_149120.1 | 2 | 3562 | ||
GenBank mRNA | AY129025.1 | - | 2 | 3568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
