siRNA Details
Gene Symbol: IL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172128.1 | NP_001165599.1 | 2 | 476 | ||
NM_172217.3 | NP_757366.2 | 1 | 216 | |||
XM_005254342.3 | XP_005254399.1 | 2 | 513 | |||
XM_011521519.2 | XP_011519821.1 | 1 | 216 | |||
XM_011521520.2 | XP_011519822.1 | 2 | 491 | |||
XM_017022140.1 | XP_016877629.1 | 1 | 216 | |||
XM_017022141.1 | XP_016877630.1 | 2 | 475 | |||
XM_017022142.1 | XP_016877631.1 | 2 | 491 | |||
XM_017022143.1 | XP_016877632.1 | 2 | 473 | |||
XM_017022144.1 | XP_016877633.1 | 2 | 490 | |||
XM_017022145.1 | XP_016877634.1 | 2 | 232 | |||
GenBank mRNA | AK124725.1 | - | 2 | 491 | ||
AK131530.1 | - | 2 | 475 | |||
AK308440.1 | - | 2 | 366 | |||
AL833554.1 | - | 4 | 382 | |||
AY324389.1 | - | 1 | 115 | |||
BC040272.1 | - | 2 | 361 | |||
BC136660.1 | - | 2 | 241 | 9020650 | ||
CR749286.1 | - | 3 | 304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
