siRNA Details
Gene Symbol: ISY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199469.1 | NP_001186398.1 | 9 | 787 | ||
NM_020701.3 | NP_065752.1 | 8 | 721 | |||
GenBank mRNA | BC004442.1 | - | 8 | 536 | 2820683 | |
BC007409.2 | - | 9 | 605 | 3030303 | ||
BC019849.1 | - | 8 | 677 | 4521880 | ||
BG282120.1 | - | 8 | 721 |
Gene Symbol: ISY1-RAB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204890.1 | NP_001191819.1 | 8 | 721 | ||
GenBank mRNA | AB032986.1 | - | 8 | 527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
