siRNA Details
Gene Symbol: ANKRD26P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027248.3 | 14 | 2426 | |||
GenBank mRNA | BC036210.1 | - | 14 | 2426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
