siRNA Details
Gene Symbol: TENM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098816.2 | NP_001092286.2 | 15 | 2575 | ||
XM_017017525.1 | XP_016873014.1 | 14 | 2846 | |||
XM_017017526.1 | XP_016873015.1 | 12 | 3751 | |||
GenBank mRNA | EG328205.1 | - | 2 | 153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
