siRNA Details
Gene Symbol: VWA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173615.4 | NP_775886.3 | 26 | 2762 | ||
XM_011545742.2 | XP_011544044.1 | 29 | 2972 | |||
XM_011545744.2 | XP_011544046.1 | 14 | 1572 | |||
XM_011545745.1 | XP_011544047.1 | 12 | 1340 | |||
XM_011545746.2 | XP_011544048.1 | 12 | 1443 | |||
XM_017022947.1 | XP_016878436.1 | 30 | 3045 | |||
XM_017022948.1 | XP_016878437.1 | 13 | 2144 | |||
XM_017022949.1 | XP_016878438.1 | 11 | 2415 | |||
GenBank mRNA | AK128606.1 | - | 15 | 1630 | ||
BC109294.1 | - | 15 | 1537 | |||
BC109295.1 | - | 14 | 1490 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
