siRNA Details
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026052.1 | 1 | 161 | |||
GenBank mRNA | AK000827.1 | - | 1 | 98 | ||
AK311347.1 | - | 1 | 98 | |||
AL708489.1 | - | 1 | 162 | |||
BC000766.2 | - | 1 | 70 | |||
BC001952.1 | - | 1 | 70 | |||
BC043401.1 | - | 1 | 100 | |||
BC065480.1 | - | 1 | 115 | |||
BC067232.1 | - | 1 | 105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
