siRNA Details
Gene Symbol: CHD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308319.1 | NP_001295248.1 | 1 | 171 | ||
NM_025134.4 | NP_079410.4 | 2 | 216 | |||
XM_005256168.3 | XP_005256225.1 | 2 | 258 | |||
XM_005256169.2 | XP_005256226.1 | 2 | 280 | |||
XM_005256172.3 | XP_005256229.1 | 2 | 197 | |||
XM_006721281.2 | XP_006721344.1 | 2 | 197 | |||
XM_006721283.3 | XP_006721346.1 | 2 | 418 | |||
XM_011523347.2 | XP_011521649.1 | 3 | 330 | |||
XM_017023718.1 | XP_016879207.1 | 3 | 453 | |||
XM_017023719.1 | XP_016879208.1 | 4 | 404 | |||
XM_017023720.1 | XP_016879209.1 | 3 | 298 | |||
XM_017023721.1 | XP_016879210.1 | 3 | 454 | |||
XM_017023722.1 | XP_016879211.1 | 3 | 454 | |||
XM_017023723.1 | XP_016879212.1 | 3 | 454 | |||
XM_017023724.1 | XP_016879213.1 | 2 | 197 | |||
XM_017023725.1 | XP_016879214.1 | 2 | 197 | |||
XM_017023728.1 | XP_016879217.1 | 2 | 256 | |||
GenBank mRNA | AK092208.1 | - | 2 | 280 | ||
AY243500.1 | - | 1 | 94 | |||
BC033770.1 | - | 2 | 226 | |||
BC140815.1 | - | 1 | 48 | 9021683 | ||
BC144622.1 | - | 1 | 48 | 9053153 | ||
DQ333316.1 | - | 2 | 310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
