siRNA Details
Gene Symbol: CCDC71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022903.3 | NP_075054.3 | 2 | 1245 | ||
GenBank mRNA | AK022862.1 | - | 2 | 1216 | ||
AK023691.1 | - | 1 | 500 | |||
AK098658.1 | - | 3 | 1080 | |||
BC035516.1 | - | 2 | 1214 | 5229681 | ||
BC071960.1 | - | 1 | 404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
