siRNA Details
Gene Symbol: TBL1XR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321193.1 | NP_001308122.1 | 14 | 1578 | ||
NM_001321194.1 | NP_001308123.1 | 15 | 1758 | |||
NM_001321195.1 | NP_001308124.1 | 13 | 1475 | |||
NM_024665.5 | NP_078941.2 | 14 | 1639 | |||
XM_005247775.2 | XP_005247832.1 | 13 | 1594 | |||
XM_006713746.1 | XP_006713809.1 | 15 | 1648 | |||
XM_011513141.1 | XP_011511443.1 | 15 | 1582 | |||
XM_011513142.2 | XP_011511444.1 | 14 | 1563 | |||
XM_011513143.2 | XP_011511445.1 | 13 | 1489 | |||
XM_017007185.1 | XP_016862674.1 | 14 | 1576 | |||
GenBank mRNA | AF268193.1 | - | 14 | 1472 | ||
AF314544.1 | - | 13 | 1438 | |||
AK022956.1 | - | 14 | 1538 | |||
AK296946.1 | - | 12 | 1220 | |||
BX648935.1 | - | 14 | 1573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
