siRNA Details
Gene Symbol: DDX47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016355.3 | NP_057439.2 | 5, 6 | 585 | ||
NM_201224.1 | NP_957518.1 | 5, 6 | 585 | |||
GenBank mRNA | AF078843.1 | - | 5, 6 | 542 | ||
AF190165.1 | - | 5, 6 | 549 | |||
AJ276704.1 | - | 5, 6 | 563 | |||
AK054574.1 | - | 5, 6 | 570 | |||
AK222940.1 | - | 5, 6 | 544 | |||
AK225144.1 | - | 5, 6 | 555 | |||
AK310501.1 | - | 5, 6 | 615 | |||
AL136666.1 | - | 5, 6 | 589 | |||
AL833469.1 | - | 4, 5 | 385 | |||
BC009379.2 | - | 5, 6 | 548 | |||
BC068009.1 | - | 5, 6 | 567 | 6142837 | ||
EF036507.1 | - | 5, 6 | 532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
