siRNA Details
Gene Symbol: DDX47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016355.3 | NP_057439.2 | 9, 10 | 1055 | ||
NM_201224.1 | NP_957518.1 | 8, 9 | 908 | |||
GenBank mRNA | AF078843.1 | - | 8, 9 | 865 | ||
AF190165.1 | - | 8, 9 | 1568 | |||
AJ276704.1 | - | 9, 10 | 1033 | |||
AK054574.1 | - | 9, 10 | 1040 | |||
AK222940.1 | - | 9, 10 | 1014 | |||
AK225144.1 | - | 9, 10 | 1025 | |||
AL136666.1 | - | 9, 10 | 1059 | |||
AL833469.1 | - | 6, 7 | 1928 | |||
BC009379.2 | - | 9, 10 | 1018 | |||
BC068009.1 | - | 9, 10 | 1037 | 6142837 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
