siRNA Details
Gene Symbol: DDX47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016355.3 | NP_057439.2 | 11 | 1209 | ||
NM_201224.1 | NP_957518.1 | 10 | 1062 | |||
GenBank mRNA | AF078843.1 | - | 10 | 1019 | ||
AF190165.1 | - | 10 | 1722 | |||
AJ276704.1 | - | 11 | 1187 | |||
AK054574.1 | - | 11 | 1194 | |||
AK222940.1 | - | 11 | 1168 | |||
AK225144.1 | - | 11 | 1179 | |||
AL136666.1 | - | 11 | 1213 | |||
AL833469.1 | - | 8 | 2082 | |||
BC009379.2 | - | 11 | 1172 | |||
BC068009.1 | - | 11 | 1191 | 6142837 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
