siRNA Details
Gene Symbol: SPTY2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194285.2 | NP_919261.2 | 3 | 1139 | ||
XM_011519919.2 | XP_011518221.1 | 3 | 997 | |||
GenBank mRNA | AF452716.1 | - | 1 | 533 | ||
AK096760.1 | - | 3 | 1159 | |||
AK304078.1 | - | 3 | 1055 | |||
BC056261.1 | - | 3 | 1014 | 6616686 | ||
BX641102.1 | - | 3 | 1040 | |||
BX647798.1 | - | 3 | 1139 | |||
BX648114.1 | - | 1 | 718 | |||
BX648478.1 | - | 3 | 1061 | |||
CR749626.1 | - | 1 | 582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
