siRNA Details
Gene Symbol: LYNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023946.3 | NP_076435.1 | 5 | 558 | ||
NM_177458.1 | NP_803253.1 | 3 | 224 | |||
GenBank mRNA | AB081838.1 | - | 3 | 224 | ||
AY358417.1 | - | 3 | 240 | |||
AY587277.1 | - | 3 | 202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
