siRNA Details
Gene Symbol: TP53TG3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006721118.1 | XP_006721181.1 | 3 | 851 | ||
XM_011546033.1 | XP_011544335.1 | 3 | 843 | |||
XM_017023955.1 | XP_016879444.1 | 3 | 3491 | |||
XM_017023956.1 | XP_016879445.1 | 3 | 3483 | |||
XM_017023957.1 | XP_016879446.1 | 4 | 1147 | |||
XM_017023958.1 | XP_016879447.1 | 4 | 1139 | |||
XM_017023959.1 | XP_016879448.1 | 2 | 3203 |
Gene Symbol: LOC102723655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724905.3 | XP_006724968.1 | 2 | 3203 | ||
XM_017030123.1 | XP_016885612.1 | 3 | 3491 | |||
XM_017030124.1 | XP_016885613.1 | 3 | 3483 |
Gene Symbol: LOC102723713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546237.1 | XP_011544539.1 | 3 | 1189 | ||
XM_011546238.1 | XP_011544540.1 | 3 | 1181 | |||
XM_017030126.1 | XP_016885615.1 | 4 | 1479 | |||
XM_017030127.1 | XP_016885616.1 | 4 | 1471 |
Gene Symbol: TP53TG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016212.4 | NP_057296.1 | 2 | 710 | ||
NR_110886.1 | 4 | 1166 | ||||
NR_110910.1 | 3 | 999 | ||||
XM_006721026.1 | XP_006721089.1 | 3 | 851 | |||
XM_011545779.1 | XP_011544081.1 | 3 | 843 | |||
XM_017023102.1 | XP_016878591.1 | 3 | 3483 | |||
XM_017023103.1 | XP_016878592.1 | 4 | 1139 | |||
GenBank mRNA | AB023508.1 | - | 4 | 1166 | ||
AK097435.1 | - | 1 | 1402 | |||
AL133588.1 | - | 2 | 1313 | |||
BC096733.1 | - | 2 | 1085 | 40015196 | ||
BC098165.1 | - | 2 | 708 | 40015195 | ||
BX381390.2 | - | 3 | 893 |
Gene Symbol: TP53TG3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099687.3 | NP_001093157.1 | 2 | 710 | ||
NR_110897.1 | 4 | 1166 | ||||
NR_110911.1 | 3 | 999 | ||||
XM_006721084.1 | XP_006721147.1 | 3 | 851 | |||
XM_011545933.1 | XP_011544235.1 | 3 | 843 | |||
XM_017023638.1 | XP_016879127.1 | 3 | 3483 | |||
XM_017023639.1 | XP_016879128.1 | 4 | 1139 |
Gene Symbol: TP53TG3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205259.2 | NP_001192188.1 | 2 | 708 | ||
NR_110914.1 | 3 | 997 | ||||
XM_006721074.2 | XP_006721137.1 | 3 | 851 | |||
XM_011545920.1 | XP_011544222.1 | 3 | 843 | |||
XM_017023591.1 | XP_016879080.1 | 3 | 3483 | |||
XM_017023592.1 | XP_016879081.1 | 4 | 1147 | |||
XM_017023593.1 | XP_016879082.1 | 4 | 1139 | |||
GenBank mRNA | DT215671.1 | - | 2 | 541 |
Gene Symbol: TP53TG3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243722.2 | NP_001230651.1 | 2 | 708 | ||
NR_110898.1 | 3 | 874 | ||||
XM_017023633.1 | XP_016879122.1 | 3 | 2395 | |||
XM_017023634.1 | XP_016879123.1 | 3 | 2387 | |||
XM_017023635.1 | XP_016879124.1 | 4 | 2563 | |||
XM_017023636.1 | XP_016879125.1 | 3 | 2273 | |||
XM_017023637.1 | XP_016879126.1 | 3 | 2265 | |||
GenBank mRNA | AB023509.1 | - | 2 | 709 | ||
BC096700.1 | - | 2 | 708 | 40015193 | ||
BC099842.1 | - | 2 | 708 | 40015192 |
Gene Symbol: TP53TG3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006721117.2 | XP_006721180.1 | 3 | 851 | ||
XM_011546032.1 | XP_011544334.1 | 3 | 843 | |||
XM_017023950.1 | XP_016879439.1 | 3 | 3491 | |||
XM_017023951.1 | XP_016879440.1 | 3 | 3483 | |||
XM_017023952.1 | XP_016879441.1 | 4 | 1147 | |||
XM_017023953.1 | XP_016879442.1 | 4 | 1139 | |||
XM_017023954.1 | XP_016879443.1 | 2 | 3203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
