siRNA Details
Gene Symbol: RNF213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256071.2 | NP_001243000.2 | 6 | 1133 | ||
NM_020954.3 | NP_066005.2 | 6 | 1133 | |||
XM_005257545.4 | XP_005257602.2 | 7 | 1321 | |||
XM_005257546.4 | XP_005257603.2 | 7 | 1225 | |||
XM_006721995.3 | XP_006722058.1 | 7 | 1608 | |||
XM_011525084.2 | XP_011523386.1 | 7 | 1321 | |||
XM_011525086.2 | XP_011523388.1 | 7 | 1321 | |||
XM_011525087.2 | XP_011523389.1 | 7 | 1321 | |||
XM_017024905.1 | XP_016880394.1 | 2 | 188 | |||
GenBank mRNA | AB046838.1 | - | 3 | 578 | ||
AB537889.1 | - | 6 | 1126 | |||
AL832920.1 | - | 6 | 1120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
