siRNA Details
Gene Symbol: ZFP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL831951.1 | - | 3 | 2315 |
Gene Symbol: GLI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138465.3 | NP_612474.1 | 3 | 233 | ||
GenBank mRNA | AB209654.1 | - | 3 | 5220 | ||
AY129020.1 | - | 1 | 129 | |||
BC014165.2 | - | 2 | 199 | 4549240 | ||
BI761835.1 | - | 3 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
