siRNA Details
Gene Symbol: ZFP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL831951.1 | - | 4 | 2643 |
Gene Symbol: GLI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138465.3 | NP_612474.1 | 4 | 561 | ||
GenBank mRNA | AY129020.1 | - | 1 | 1551 | ||
BC014165.2 | - | 3 | 527 | 4549240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
