siRNA Details
Gene Symbol: TNRC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080495.2 | NP_001073964.2 | 11 | 4200 | ||
XM_017012728.1 | XP_016868217.1 | 11 | 6697 | |||
XM_017012729.1 | XP_016868218.1 | 10 | 4404 | |||
XM_017012730.1 | XP_016868219.1 | 10 | 4019 | |||
XM_017012731.1 | XP_016868220.1 | 10 | 4023 | |||
XM_017012732.1 | XP_016868221.1 | 10 | 4020 | |||
XM_017012733.1 | XP_016868222.1 | 9 | 3879 | |||
XM_017012734.1 | XP_016868223.1 | 10 | 4905 | |||
XM_017012735.1 | XP_016868224.1 | 10 | 3907 | |||
XM_017012736.1 | XP_016868225.1 | 10 | 4494 | |||
XM_017012737.1 | XP_016868226.1 | 10 | 6267 | |||
XM_017012738.1 | XP_016868227.1 | 10 | 3789 | |||
XM_017012739.1 | XP_016868228.1 | 10 | 4024 | |||
XM_017012740.1 | XP_016868229.1 | 10 | 4024 | |||
GenBank mRNA | AB058759.1 | - | 1 | 485 | ||
AK027341.1 | - | 2 | 660 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
