siRNA Details
Gene Symbol: ERCC6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020207.4 | NP_064592.2 | 17 | 3856 | ||
XM_011518641.2 | XP_011516943.1 | 16 | 3703 | |||
XM_011518644.2 | XP_011516946.1 | 16 | 3648 | |||
XM_011518645.2 | XP_011516947.1 | 17 | 3856 | |||
XM_011518646.2 | XP_011516948.1 | 17 | 3856 | |||
XM_011518647.1 | XP_011516949.1 | 17 | 3856 | |||
XM_011518648.2 | XP_011516950.1 | 8 | 1928 | |||
XM_011518650.2 | XP_011516952.1 | 17 | 3175 | |||
XM_017014707.1 | XP_016870196.1 | 17 | 3843 | |||
GenBank mRNA | AK308131.1 | - | 2 | 891 | ||
AL133663.1 | - | 12 | 2456 | |||
BC022957.1 | - | 2 | 285 | 4995030 | ||
BG108141.1 | - | 1 | 116 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
