siRNA Details
Gene Symbol: PRAMEF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012276.3 | NP_001012276.2 | 3 | 872 | ||
XM_017001269.1 | XP_016856758.1 | 2 | 868 | |||
GenBank mRNA | BC140755.1 | - | 3 | 872 | 9021623 | |
BC144565.1 | - | 2 | 874 | 9053093 |
Gene Symbol: PRAMEF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012277.4 | NP_001012277.2 | 3 | 872 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
