siRNA Details
Gene Symbol: SPATA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166271.2 | NP_001159743.1 | 7 | 2976 | ||
NM_001286792.1 | NP_001273721.1 | 9 | 3300 | |||
NM_001286794.1 | NP_001273723.1 | 5 | 918 | |||
NM_001286795.1 | NP_001273724.1 | 4 | 710 | |||
NM_153023.3 | NP_694568.1 | 6 | 1212 | |||
GenBank mRNA | AK055770.1 | - | 6 | 1180 | ||
AK123031.1 | - | 4 | 610 | |||
AK160371.1 | - | 3 | 399 | |||
AK298717.1 | - | 5 | 820 | |||
AK299981.1 | - | 4 | 653 | |||
BC109290.1 | - | 5 | 922 | 40034357 | ||
BC109291.1 | - | 5 | 922 | 40034359 | ||
BX648244.1 | - | 9 | 3122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
