siRNA Details
Gene Symbol: SPATA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166271.2 | NP_001159743.1 | 8 | 3150 | ||
NM_001286792.1 | NP_001273721.1 | 10 | 3474 | |||
NM_001286793.1 | NP_001273722.1 | 4 | 698 | |||
NM_001286794.1 | NP_001273723.1 | 6 | 1092 | |||
NM_001286795.1 | NP_001273724.1 | 5 | 884 | |||
NM_153023.3 | NP_694568.1 | 7 | 1386 | |||
GenBank mRNA | AK055770.1 | - | 7 | 1354 | ||
AK097217.1 | - | 1 | 27 | |||
AK123031.1 | - | 5 | 784 | |||
AK160371.1 | - | 4 | 573 | |||
AK297394.1 | - | 4 | 668 | |||
AK298717.1 | - | 6 | 994 | |||
AK299981.1 | - | 5 | 827 | |||
AK301149.1 | - | 4 | 641 | |||
AK307528.1 | - | 1 | 103 | |||
BC109290.1 | - | 6 | 1096 | 40034357 | ||
BC109291.1 | - | 6 | 1096 | 40034359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
