siRNA Details
Gene Symbol: EP400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015409.4 | NP_056224.3 | 11 | 2869 | ||
GenBank mRNA | AB040931.1 | - | 10 | 2722 | ||
AY044869.1 | - | 10 | 2753 | |||
BC037208.1 | - | 12 | 2926 | |||
BC150291.1 | - | 10 | 2722 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
