siRNA Details
Gene Symbol: EP400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015409.4 | NP_056224.3 | 14 | 3147 | ||
GenBank mRNA | AB040931.1 | - | 13 | 3000 | ||
AY044869.1 | - | 13 | 3031 | |||
BC150291.1 | - | 13 | 3000 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
