siRNA Details
Gene Symbol: NKRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ011812.2 | - | 2 | 872 | ||
EF158333.1 | - | 2 | 872 | |||
EF165090.1 | - | 2 | 612 | |||
EF165091.1 | - | 2 | 691 | |||
EF165093.1 | - | 2 | 762 | |||
EF216688.1 | - | 2 | 872 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
