siRNA Details
Gene Symbol: LRTOMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145307.4 | NP_001138779.1 | 5 | 1094 | ||
NM_001145308.4 | NP_001138780.1 | 3 | 873 | |||
NM_001145309.3 | NP_001138781.1 | 5 | 1094 | |||
NM_001145310.3 | NP_001138782.1 | 5 | 1094 | |||
NM_001205138.3 | NP_001192067.1 | 4 | 957 | |||
NM_001271471.2 | NP_001258400.1 | 5 | 1094 | |||
NM_001318803.1 | NP_001305732.1 | 5 | 1051 | |||
NM_145309.5 | NP_660352.1 | 5 | 1094 | |||
NR_026886.3 | 6 | 1274 | ||||
NR_134858.1 | 3 | 873 | ||||
XM_006718473.3 | XP_006718536.1 | 5 | 1320 | |||
XM_006718474.3 | XP_006718537.1 | 4 | 1359 | |||
XM_011544847.2 | XP_011543149.1 | 5 | 1548 | |||
XM_011544848.2 | XP_011543150.1 | 6 | 1606 | |||
XM_017017359.1 | XP_016872848.1 | 6 | 1711 | |||
XM_017017360.1 | XP_016872849.1 | 6 | 1563 | |||
GenBank mRNA | AF289588.1 | - | 1 | 76 | ||
AK298997.1 | - | 4 | 476 | |||
AK302772.1 | - | 3 | 367 | |||
AK311690.1 | - | 6 | 744 | |||
AK313150.1 | - | 5 | 778 | |||
AK315892.1 | - | 2 | 172 | |||
BC012855.1 | - | 5 | 566 | 3863323 | ||
BI461207.1 | - | 5 | 537 | |||
DA004114.1 | - | 3 | 427 | |||
EU627066.1 | - | 5 | 566 | |||
EU627067.1 | - | 4 | 516 | |||
EU627068.1 | - | 5 | 566 | |||
EU627069.1 | - | 5 | 566 | |||
EU627070.1 | - | 5 | 566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
