siRNA Details
Gene Symbol: LRTOMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145307.4 | NP_001138779.1 | 2 | 614 | ||
NM_001145309.3 | NP_001138781.1 | 2 | 614 | |||
NM_001145310.3 | NP_001138782.1 | 2 | 614 | |||
NM_001205138.3 | NP_001192067.1 | 2 | 614 | |||
NM_001271471.2 | NP_001258400.1 | 2 | 614 | |||
NM_001318803.1 | NP_001305732.1 | 2 | 571 | |||
NM_145309.5 | NP_660352.1 | 2 | 614 | |||
NR_026886.3 | 2 | 614 | ||||
XM_006718473.3 | XP_006718536.1 | 2 | 840 | |||
XM_011544847.2 | XP_011543149.1 | 2 | 1068 | |||
XM_011544848.2 | XP_011543150.1 | 3 | 1126 | |||
XM_017017359.1 | XP_016872848.1 | 3 | 1231 | |||
XM_017017360.1 | XP_016872849.1 | 3 | 1083 | |||
GenBank mRNA | AK298997.1 | - | 2 | 133 | ||
AK311690.1 | - | 2 | 84 | |||
AK313150.1 | - | 2 | 298 | |||
BC012855.1 | - | 2 | 86 | 3863323 | ||
BI461207.1 | - | 2 | 57 | |||
EU627066.1 | - | 2 | 86 | |||
EU627067.1 | - | 1 | 36 | |||
EU627068.1 | - | 2 | 86 | |||
EU627069.1 | - | 2 | 86 | |||
EU627070.1 | - | 2 | 86 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
