siRNA Details
Gene Symbol: MROH7-TTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037639.2 | 3 | 1317 | |||
NR_037640.2 | 3 | 1212 | ||||
NR_037641.2 | 3 | 1212 | ||||
GenBank mRNA | BX648088.1 | - | 3 | 1150 | ||
CR749830.1 | - | 3 | 1269 |
Gene Symbol: MROH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039464.3 | NP_001034553.2 | 3 | 1212 | ||
NR_026782.2 | 3 | 1206 | ||||
NR_111931.1 | 3 | 1212 | ||||
GenBank mRNA | AK093272.1 | - | 3 | 1151 | ||
AK130047.1 | - | 2 | 1701 | |||
AK302446.1 | - | 3 | 1159 | |||
AL832492.1 | - | 1 | 439 | |||
BC095478.1 | - | 2 | 994 | |||
BC126126.1 | - | 1 | 912 | 8991842 | ||
BC143891.1 | - | 1 | 912 | 9052408 | ||
BC144516.1 | - | 3 | 1150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
