siRNA Details
Gene Symbol: RBM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053043.2 | NP_444271.2 | 6 | 1076 | ||
XM_005249531.1 | XP_005249588.1 | 7 | 1211 | |||
XM_005249532.1 | XP_005249589.1 | 7 | 1211 | |||
XM_005249533.1 | XP_005249590.1 | 7 | 1211 | |||
XM_005249534.1 | XP_005249591.1 | 6 | 1091 | |||
XM_005249535.1 | XP_005249592.1 | 6 | 1091 | |||
XM_011515840.1 | XP_011514142.1 | 7 | 1211 | |||
GenBank mRNA | AK301179.1 | - | 1 | 23 | ||
AK307515.1 | - | 6 | 906 | |||
AL137724.1 | - | 4 | 573 | |||
BC011923.2 | - | 6 | 1134 | |||
DA947894.1 | - | 1 | 23 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
