siRNA Details
Gene Symbol: UBOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267584.1 | NP_001254513.1 | 5 | 3283 | ||
NM_014948.3 | NP_055763.1 | 5 | 3288 | |||
NM_199415.2 | NP_955447.1 | 4 | 3126 | |||
GenBank mRNA | AB020667.1 | - | 5 | 3257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
