siRNA Details
Gene Symbol: UBOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267584.1 | NP_001254513.1 | 2 | 219 | ||
NM_014948.3 | NP_055763.1 | 2 | 219 | |||
NM_199415.2 | NP_955447.1 | 2 | 219 | |||
GenBank mRNA | AB020667.1 | - | 2 | 187 | ||
AK022444.1 | - | 2 | 183 | |||
AK222570.1 | - | 2 | 191 | |||
AK222876.1 | - | 2 | 144 | |||
BC000515.2 | - | 2 | 139 | 2822816 | ||
BC046122.1 | - | 2 | 179 | 5554044 | ||
BX475562.1 | - | 2 | 508 | |||
CR457089.1 | - | 1 | 33 | |||
DA101925.1 | - | 2 | 219 |

More Information
Other Species Targeted

Gene Ontology Categories:
