siRNA Details
Gene Symbol: BRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242786.1 | NP_001229715.1 | 7, 8 | 866 | ||
NM_001242787.1 | NP_001229716.1 | 7, 8 | 866 | |||
NM_001242788.1 | NP_001229717.1 | 6, 7 | 1076 | |||
NM_001242789.1 | NP_001229718.1 | 2, 3 | 356 | |||
NM_001519.3 | NP_001510.2 | 7, 8 | 1157 | |||
NM_145685.2 | NP_663718.1 | 3, 4 | 1065 | |||
XM_005267561.3 | XP_005267618.1 | 7, 8 | 1179 | |||
XM_005267563.3 | XP_005267620.1 | 3, 4 | 1869 | |||
XM_006720123.2 | XP_006720186.1 | 2, 3 | 1297 | |||
XM_011536672.2 | XP_011534974.1 | 7, 8 | 1179 | |||
XM_011536673.1 | XP_011534975.1 | 6, 7 | 1029 | |||
XM_011536674.1 | XP_011534976.1 | 3, 4 | 1447 | |||
GenBank mRNA | AJ297406.1 | - | 3, 4 | 1619 | ||
AJ297407.1 | - | 3, 4 | 1073 | |||
AK096471.1 | - | 2, 3 | 356 | |||
AK294899.1 | - | 7, 8 | 866 | |||
AK295579.1 | - | 7, 8 | 779 | |||
BC071637.1 | - | 2, 3 | 221 | |||
BC086856.1 | - | 6, 7 | 809 | 6166751 | ||
BC103859.1 | - | 3, 4 | 578 | |||
BC103860.2 | - | 2, 3 | 223 | |||
BC103861.2 | - | 3, 4 | 373 | |||
BC133655.1 | - | 3, 4 | 373 | |||
U28838.1 | - | 7, 8 | 1160 | |||
U75276.1 | - | 7, 8 | 861 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
