siRNA Details
Gene Symbol: BRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242786.1 | NP_001229715.1 | 18 | 3143 | ||
NM_001242787.1 | NP_001229716.1 | 18 | 3077 | |||
NM_001242788.1 | NP_001229717.1 | 17 | 3287 | |||
NM_001242789.1 | NP_001229718.1 | 13 | 2567 | |||
NM_001519.3 | NP_001510.2 | 18 | 3368 | |||
NM_145685.2 | NP_663718.1 | 14 | 3276 | |||
XM_005267561.3 | XP_005267618.1 | 18 | 3387 | |||
XM_005267563.3 | XP_005267620.1 | 14 | 4080 | |||
XM_006720123.2 | XP_006720186.1 | 13 | 3508 | |||
XM_011536673.1 | XP_011534975.1 | 17 | 3240 | |||
XM_011536674.1 | XP_011534976.1 | 14 | 3658 | |||
GenBank mRNA | AJ297406.1 | - | 5 | 1774 | ||
AJ297407.1 | - | 14 | 3282 | |||
AK096471.1 | - | 13 | 2567 | |||
AK124665.1 | - | 8 | 2432 | |||
AK125024.1 | - | 5 | 2882 | |||
AK126499.1 | - | 1 | 3377 | |||
BC071637.1 | - | 13 | 2432 | |||
BC086856.1 | - | 17 | 3020 | 6166751 | ||
BC103859.1 | - | 5 | 733 | |||
BC103860.2 | - | 4 | 316 | |||
BC103861.2 | - | 7 | 673 | |||
BC133655.1 | - | 7 | 662 | |||
U28838.1 | - | 18 | 3358 | |||
U75276.1 | - | 18 | 3074 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
