siRNA Details
Gene Symbol: BRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242786.1 | NP_001229715.1 | 3 | 491 | ||
NM_001242787.1 | NP_001229716.1 | 3 | 491 | |||
NM_001242788.1 | NP_001229717.1 | 2 | 701 | |||
NM_001242790.1 | NP_001229719.1 | 3 | 782 | |||
NM_001519.3 | NP_001510.2 | 3 | 782 | |||
XM_005267561.3 | XP_005267618.1 | 3 | 804 | |||
XM_011536672.2 | XP_011534974.1 | 3 | 804 | |||
XM_011536673.1 | XP_011534975.1 | 3 | 804 | |||
GenBank mRNA | AJ297408.1 | - | 3 | 361 | ||
AK294899.1 | - | 3 | 491 | |||
AK295579.1 | - | 3 | 404 | |||
BC016743.2 | - | 3 | 728 | |||
BC086856.1 | - | 2 | 434 | 6166751 | ||
EU668327.1 | - | 3 | 728 | |||
U28838.1 | - | 3 | 785 | |||
U75276.1 | - | 3 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
