siRNA Details
Gene Symbol: SP140L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308162.1 | NP_001295091.1 | 12 | 1143 | ||
NM_001308163.1 | NP_001295092.1 | 11 | 1038 | |||
NM_138402.4 | NP_612411.4 | 13 | 1219 | |||
XM_006712855.2 | XP_006712918.1 | 12 | 1100 | |||
XM_006712856.2 | XP_006712919.1 | 12 | 1100 | |||
XM_006712858.3 | XP_006712921.1 | 11 | 1064 | |||
XM_011512190.2 | XP_011510492.1 | 13 | 1256 | |||
XM_017005294.1 | XP_016860783.1 | 13 | 1256 | |||
XM_017005295.1 | XP_016860784.1 | 13 | 1256 | |||
XM_017005296.1 | XP_016860785.1 | 13 | 1205 | |||
XM_017005297.1 | XP_016860786.1 | 12 | 1222 | |||
XM_017005298.1 | XP_016860787.1 | 12 | 1151 | |||
XM_017005299.1 | XP_016860788.1 | 12 | 1170 | |||
XM_017005300.1 | XP_016860789.1 | 13 | 1205 | |||
GenBank mRNA | AK023116.1 | - | 8 | 616 | ||
BC105007.1 | - | 7 | 485 | |||
BC143624.1 | - | 7 | 485 | |||
BQ232706.1 | - | 7 | 539 | |||
CR987282.1 | - | 4 | 390 | |||
KF419365.1 | - | 12 | 1053 | |||
KF419366.1 | - | 11 | 948 | |||
KF419367.1 | - | 11 | 948 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
