siRNA Details
Gene Symbol: ARL17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113738.1 | NP_001107210.1 | 4 | 2741 | ||
GenBank mRNA | AB209344.1 | - | 4 | 2672 | ||
BX537922.1 | - | 1 | 1540 | |||
BX647266.1 | - | 4 | 2701 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
