siRNA Details
Gene Symbol: ZNF729 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242680.1 | NP_001229609.1 | 4 | 1588 | ||
XM_017026122.1 | XP_016881611.1 | 4 | 1634 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
